ENA161669295 Enamine

1-(2-amino-3-methylbenzoyl)piperidin-4-ol

  • MDL number MFCD09729491
  • Cas Number : 953735-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36538
Availability details for
Quantity
  • Availability
  • Add to Cart