ENA161669304 Enamine

1H-benzimidazole-2-sulfonic acid

  • MDL number MFCD00100972
  • Cas Number : 40828-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Danger
safety_statement 26-36/37/39-45
msds https://www.enaminestore.com/catalog/EN300-36560
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart