ENA161669307 Enamine

1-(3-methylbutyl)-1H-benzimidazole-2-sulfonic acid

  • MDL number MFCD03670848
  • Cas Number : 537009-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36563
Availability details for
Quantity
  • Availability
  • Add to Cart