ENA161669317 Enamine

N-[1-(1H-benzimidazol-2-yl)ethyl]-2-furamide

  • MDL number MFCD02982194
  • Cas Number : 376620-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36573
Availability details for
Quantity
  • Availability
  • Add to Cart