ENA161669318 Enamine

N-[3-(1H-benzimidazol-2-yl)propyl]-2-methylpropanamide

  • MDL number MFCD04218420
  • Cas Number : 637322-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36574
Availability details for
Quantity
  • Availability
  • Add to Cart