ENA161669321 Enamine

2-chloro-4-fluoro-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD10686916
  • Cas Number : 125607-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36580
Availability details for
Quantity
  • Availability
  • Add to Cart