ENA161669322 Enamine

2-(2,6-dimethylpiperidine-1-carbonyl)-4,5-dimethoxyaniline

  • MDL number MFCD09729555
  • Cas Number : 953892-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36582
Availability details for
Quantity
  • Availability
  • Add to Cart