ENA161669327 Enamine

1-(2-methylpropyl)piperidine-2-carboxylic acid

  • MDL number MFCD08445056
  • Cas Number : 1044637-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36587
Availability details for
Quantity
  • Availability
  • Add to Cart