ENA161669338 Enamine

2-[4-(chlorosulfonyl)phenoxy]acetic acid

  • MDL number MFCD09947771
  • Cas Number : 17641-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-147
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36605
Availability details for
Quantity
  • Availability
  • Add to Cart