ENA161669342 Enamine

1-(4-bromophenyl)propan-1-amine hydrochloride

  • MDL number MFCD10686922
  • Cas Number : 90485-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36611
Availability details for
Quantity
  • Availability
  • Add to Cart