ENA161669343 Enamine

1-(3,4-diethoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD05983789
  • Cas Number : 390815-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36612
Availability details for
Quantity
  • Availability
  • Add to Cart