ENA161669354 Enamine

2-(2-aminophenyl)-N-methylacetamide

  • MDL number MFCD09045236
  • Cas Number : 4103-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36624
Availability details for
Quantity
  • Availability
  • Add to Cart