ENA161669357 Enamine

1,1,1-trichloro-2-methylpropan-2-ol

  • MDL number MFCD00004461
  • Cas Number : 57-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-19331

Safety Information

signalword Warning
safety_statement 26-36/37/39
msds https://www.enaminestore.com/catalog/EN300-19331
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart