ENA161669358 Enamine

N-(5-amino-2-fluorophenyl)-4-methylbenzene-1-sulfonamide

  • MDL number MFCD09049316
  • Cas Number : 926255-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36629
Availability details for
Quantity
  • Availability
  • Add to Cart