ENA161669361 Enamine

5-ethyl-1-(4-nitrophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD10686929
  • Cas Number : 1152882-20-6
  • Molecular Weight : 261.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36632
Availability details for
Quantity
  • Availability
  • Add to Cart