ENA161669370 Enamine

{4-[1-(hydroxyimino)ethyl]phenyl}urea

  • Cas Number : 128369-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36656
Availability details for
Quantity
  • Availability
  • Add to Cart