ENA161669374 Enamine

6-(2,6-dimethylmorpholin-4-yl)pyridine-3-carboxylic acid

  • MDL number MFCD09047840
  • Cas Number : 926271-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36660
Availability details for
Quantity
  • Availability
  • Add to Cart