ENA161669376 Enamine

5-sulfamoylfuran-2-carboxylic acid

  • MDL number MFCD09037277
  • Cas Number : 98027-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36664
Availability details for
Quantity
  • Availability
  • Add to Cart