ENA161669383 Enamine

4-[(3,5-dichlorobenzoyl)amino]benzoic acid

  • MDL number MFCD09042734
  • Cas Number : 54057-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 305...307
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36675

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36675
Availability details for
Quantity
  • Availability
  • Add to Cart