ENA161669384 Enamine

4-[(4,4-dimethyl-2,5-dioxoimidazolidin-1-yl)methyl]benzonitrile

  • MDL number MFCD09928169
  • Cas Number : 164648-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36676
Availability details for
Quantity
  • Availability
  • Add to Cart