ENA161669389 Enamine

4-hydroxy-N,N-dimethylbenzamide

  • MDL number MFCD01168919
  • Cas Number : 20876-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36682
Availability details for
Quantity
  • Availability
  • Add to Cart