ENA161669392 Enamine

1-(2-fluorophenyl)-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09046271
  • Cas Number : 926261-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36688
Availability details for
Quantity
  • Availability
  • Add to Cart