ENA161669393 Enamine

3-hydroxy-N-methylbenzamide

  • MDL number MFCD08729289
  • Cas Number : 15788-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-115
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36689
Availability details for
Quantity
  • Availability
  • Add to Cart