ENA161669394 Enamine

2-(3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetic acid

  • MDL number MFCD09046965
  • Cas Number : 56787-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36690
Availability details for
Quantity
  • Availability
  • Add to Cart