ENA161669395 Enamine

5-[(3-ethylphenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08444648
  • Cas Number : 923153-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36693
Availability details for
Quantity
  • Availability
  • Add to Cart