ENA161669396 Enamine

2-amino-1-(4-fluorophenyl)ethanol

  • MDL number MFCD08056632
  • Cas Number : 456-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65...67
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36694

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-36694
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart