ENA161669405 Enamine

1,2-bis(4-tert-butylphenyl)-2-hydroxyethan-1-one

  • MDL number MFCD00055453
  • Cas Number : 77387-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36706
Availability details for
Quantity
  • Availability
  • Add to Cart