ENA161669416 Enamine

3-chloro-4-(difluoromethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD09804955
  • Cas Number : 1016714-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36729
Availability details for
Quantity
  • Availability
  • Add to Cart