ENA161669419 Enamine

1-(3-chlorophenyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD03161248
  • Cas Number : 143328-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36732
Availability details for
Quantity
  • Availability
  • Add to Cart