ENA161669420 Enamine

2-{[(4-bromo-3-methylphenyl)sulfonyl]amino}-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD04941580
  • Cas Number : 790284-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36733
Availability details for
Quantity
  • Availability
  • Add to Cart