ENA161669424 Enamine

4H-1,2,4-triazole-3-carbothioamide

  • MDL number MFCD09046139
  • Cas Number : 3641-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 348-350
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36766
Availability details for
Quantity
  • Availability
  • Add to Cart