ENA181911174 Enamine

N-ethyl-1-methyl-1H,2H,3H,4H-pyrrolo[1,2-a]pyrazine-6-carboxamide

  • MDL number MFCD11099401
  • Cas Number : 1087792-08-2
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31643
Availability details for
Quantity
  • Availability
  • Add to Cart