ENA181911186 Enamine

3-(1,3-benzothiazol-2-yl)propan-1-amine dihydrochloride

  • MDL number MFCD11099434
  • Cas Number : 1177309-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36832
Availability details for
Quantity
  • Availability
  • Add to Cart