ENA181911191 Enamine

1-(2-chloroacetyl)-N-(2-hydroxyphenyl)piperidine-4-carboxamide

  • MDL number MFCD11099443
  • Cas Number : 1087792-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36856
Availability details for
Quantity
  • Availability
  • Add to Cart