ENA181911193 Enamine

N-(3-chlorophenyl)-2-(methylamino)acetamide hydrochloride

  • MDL number MFCD11099450
  • Cas Number : 116133-19-8
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36882
Availability details for
Quantity
  • Availability
  • Add to Cart