ENA181911194 Enamine

N-(2-methoxyphenyl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD11099451
  • Cas Number : 1019851-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36883
Availability details for
Quantity
  • Availability
  • Add to Cart