ENA181911198 Enamine

5-bromo-4-(chlorosulfonyl)thiophene-2-carboxylic acid

  • MDL number MFCD11099459
  • Cas Number : 172461-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-36917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36917
Availability details for
Quantity
  • Availability
  • Add to Cart