ENA181911206 Enamine

1-[4-(methylthio)phenyl]ethanamine hydrochloride

  • MDL number MFCD11099481
  • Cas Number : 205937-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37028

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37028
Availability details for
Quantity
  • Availability
  • Add to Cart