ENA181911219 Enamine

3-[(trifluoromethyl)sulfanyl]propanoic acid

  • MDL number MFCD11099524
  • Cas Number : 29271-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37282
Availability details for
Quantity
  • Availability
  • Add to Cart