ENA181911228 Enamine

potassium 3-(piperazin-1-yl)propanoate

  • MDL number MFCD11099551
  • Cas Number : 1170114-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37447
Availability details for
Quantity
  • Availability
  • Add to Cart