ENA181911233 Enamine

methyl 13-oxo-12-oxa-2,3,5,7-tetraazatricyclo[7.4.0.0,2,6]trideca-1(9),3,5,7-tetraene-4-carboxylate

  • MDL number MFCD11099562
  • Cas Number : 1087792-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37525

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-37525
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart