ENA181911237 Enamine

(1,2,3-thiadiazol-4-yl)methanol

  • MDL number MFCD06202704
  • Cas Number : 50868-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37535
Availability details for
Quantity
  • Availability
  • Add to Cart