ENA181911239 Enamine

N-[(3-methoxyphenyl)methyl]-1H-imidazole-1-carboxamide

  • MDL number MFCD11099568
  • Cas Number : 1087792-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37544
Availability details for
Quantity
  • Availability
  • Add to Cart