ENA181911254 Enamine

(3-phenyl-1,2-oxazol-5-yl)methanesulfonyl chloride

  • MDL number MFCD11099587
  • Cas Number : 1087792-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38916
Availability details for
Quantity
  • Availability
  • Add to Cart