ENA181911263 Enamine

3-methyl-1-(piperidine-4-carbonyl)piperidine hydrochloride

  • MDL number MFCD11099597
  • Cas Number : 1170118-61-2
  • Molecular Weight : 246.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38963
Availability details for
Quantity
  • Availability
  • Add to Cart