ENA181911266 Enamine

ethyl 6-(3-hydroxypropyl)-2-methylpyrazolo[1,5-a]pyrimidine-7-carboxylate

  • MDL number MFCD11099599
  • Cas Number : 1087792-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38972

Safety Information

msds https://www.enaminestore.com/catalog/EN300-38972
Availability details for
Quantity
  • Availability
  • Add to Cart