ENA181911268 Enamine

2-{2-[(pyridin-2-yl)methyl]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD11099602
  • Cas Number : 1087792-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38989
Availability details for
Quantity
  • Availability
  • Add to Cart