ENA181911278 Enamine

1-sulfamoylpiperidine-4-carboxamide

  • MDL number MFCD11099618
  • Cas Number : 1087792-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39040
Availability details for
Quantity
  • Availability
  • Add to Cart