ENA181911281 Enamine

3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamide

  • MDL number MFCD11099620
  • Cas Number : 1087784-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39050
Availability details for
Quantity
  • Availability
  • Add to Cart