ENA181911289 Enamine

N-cyclopropyl-4-methylcyclohexan-1-amine hydrochloride

  • MDL number MFCD11099629
  • Cas Number : 1177271-63-4
  • Molecular Weight : 189.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39080
Availability details for
Quantity
  • Availability
  • Add to Cart